Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15C9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225251

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225251

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Applications Western Blotting
Cross Reactivity Mouse
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Regulatory Status RUO
Host Species Rat
Quantity 0.1mg
Antigen Noxa
Format Purified
Gene ID (Entrez) 5366
Clone 69N15C9
Formulation Noxa antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This product reacts with Mouse.
Concentration 0.5mg/mL
Conjugate Unlabeled