Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15C9, Novus Biologicals™

$192.00 - $406.00

Specifications

Clone 69N15C9
Host Species Rat
Isotype κ
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Antigen Noxa
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225266SS

 
novus biologicals canada
NBP225266SS
Unlabeled 0.025mg Each for $192.00

NBP225266

 
novus biologicals canada
NBP225266
Unlabeled 0.1mg Each for $406.00
Description & Specifications

Specifications

Clone 69N15C9
Host Species Rat
Isotype κ
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Antigen Noxa
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This product reacts with Mouse.
Gene ID (Entrez) 5366
Formulation Noxa antibody at 0.5mg/mL in sterile PBS
Concentration 0.5mg/mL