Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15C9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225266SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225266SS

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mg
Clone 69N15C9
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Regulatory Status RUO
Isotype κ
Antigen Noxa
Format Purified
Formulation Noxa antibody at 0.5mg/mL in sterile PBS
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Concentration 0.5mg/mL
Test Specificity This product reacts with Mouse.
Primary or Secondary Primary
Gene ID (Entrez) 5366
Cross Reactivity Mouse
Applications Western Blotting
Host Species Rat