Please login to your online account to display your discounted pricing

anti-Noxa, Clone: 69N15C9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225266

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225266

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Quantity 0.1mg
Cross Reactivity Mouse
Format Purified
Clone 69N15C9
Applications Western Blotting
Isotype κ
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Concentration 0.5mg/mL
Gene ID (Entrez) 5366
Conjugate Unlabeled
Antigen Noxa
Formulation Noxa antibody at 0.5mg/mL in sterile PBS
Immunogen Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
Test Specificity This product reacts with Mouse.
Regulatory Status RUO