Please login to your online account to display your discounted pricing

anti-MHC Class I, Clone: ER-HR52, Novus Biologicals™

$110.17 - $329.00

Specifications

Clone ER-HR52
Host Species Rat
Isotype IgG2a
Immunogen Adherent FI (CBAXBL) bone marrow stromal cells
Antigen MHC Class I
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10064952T

 
novus biologicals canada
NB10064952SS
Unlabeled 0.025mg Each for $110.17

NB10064952

 
novus biologicals canada
NB10064952
Unlabeled 0.125mg Each for $329.00
Description & Specifications

Specifications

Clone ER-HR52
Host Species Rat
Isotype IgG2a
Immunogen Adherent FI (CBAXBL) bone marrow stromal cells
Antigen MHC Class I
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Test Specificity NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2 d,p,q. Mouse strains with the haplotypes H-2 f,r,s,w17,w23,w27 show weak reactivity with this antibody.
Gene ID (Entrez) 3133
Formulation Phosphate buffered saline pH 7.4
Concentration 1.0mg/mL