Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Clone: 73N4E10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225246

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225246

  • / Each
 

Description & Specifications

Specifications

Antigen IL-7 R alpha/CD127
Applications Western Blot
Clone 73N4E10
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Purified
Formulation IL-7R/CD127 antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Host Species Rat
Immunogen Amino acids 145-235 of mouse IL-7R/CD127 were used as the immunogen for the antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3575
Test Specificity This product reacts with Mouse.
Target Species Murine