Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Clone: 73N4E10, Novus Biologicals™

$109.00 - $109.00

Specifications

Clone 73N4E10
Host Species Rat
Isotype κ
Immunogen Amino acids 145-235 of mouse IL-7R/CD127 were used as the immunogen for the antibody.
Antigen IL-7 R alpha/CD127
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225246SS

 
novus biologicals canada
NBP225246SS
Unlabeled 0.025mg Each for $109.00

NBP225246

 
novus biologicals canada
NBP225246
Unlabeled 0.1mg Each for $109.00
Description & Specifications

Specifications

Clone 73N4E10
Host Species Rat
Isotype κ
Immunogen Amino acids 145-235 of mouse IL-7R/CD127 were used as the immunogen for the antibody.
Antigen IL-7 R alpha/CD127
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This product reacts with Mouse.
Gene ID (Entrez) 3575
Formulation IL-7R/CD127 antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Concentration 0.5mg/mL