Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Clone: 73N4E10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225263

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225263

  • / Each
 

Description & Specifications

Specifications

Format Purified
Quantity 0.1mg
Regulatory Status RUO
Host Species Rat
Monoclonal or Polyclonal Monoclonal
Test Specificity This product reacts with Mouse.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Formulation IL-7R/CD127 antibody at 0.5mg/mL in sterile PBS.
Applications Western Blotting
Immunogen Amino acids 145-235 of mouse IL-7R/CD127 were used as the immunogen for the antibody.
Isotype κ
Primary or Secondary Primary
Gene ID (Entrez) 3575
Clone 73N4E10
Antigen IL-7 R alpha/CD127