Please login to your online account to display your discounted pricing

anti-IL-7 R alpha/CD127, Clone: 73N4E10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225246SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225246SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Test Specificity This product reacts with Mouse.
Conjugate Unlabeled
Concentration 0.5mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 3575
Formulation IL-7R/CD127 antibody at 0.5mg/mL in PBS containing, 0.05% BSA
Isotype κ
Immunogen Amino acids 145-235 of mouse IL-7R/CD127 were used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Cross Reactivity Mouse
Host Species Rat
Antigen IL-7 R alpha/CD127
Applications Western Blotting
Format Purified
Clone 73N4E10