Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 5P6G8, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 5P6G8
Host Species Rat
Isotype κ
Immunogen Full-length recombinant mouse EBI3 protein
Antigen IL-27/IL-35 EBI3 Subunit
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203942

 
novus biologicals canada
NBP203942
Unlabeled 0.1mg Each for $329.00

NBP203942SS

 
novus biologicals canada
NBP203942SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 5P6G8
Host Species Rat
Isotype κ
Immunogen Full-length recombinant mouse EBI3 protein
Antigen IL-27/IL-35 EBI3 Subunit
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Western Blot
Gene ID (Entrez) 10148
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 1.0mg/mL