Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 10J811, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203941SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203941SS

  • / Each
 

Description & Specifications

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Applications Western Blot
Clone 10J811
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Host Species Rat
Immunogen Full-length recombinant mouse EBI3 protein
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10148
Target Species Murine