Please login to your online account to display your discounted pricing

anti-IL-27/IL-35 EBI3 Subunit, Clone: 10J803, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203940

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203940

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Gene ID (Entrez) 10148
Monoclonal or Polyclonal Monoclonal
Immunogen Full-length recombinant mouse EBI3 protein
Isotype κ
Quantity 0.1mg
Cross Reactivity Mouse
Format Purified
Regulatory Status RUO
Host Species Rat
Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Applications Western Blotting
Clone 10J803