Please login to your online account to display your discounted pricing

anti-GM-CSF, Clone: BVD2-21C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232967

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232967

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Quantity 0.5mL
Gene Accession No. P04141
Conjugate Unlabeled
Regulatory Status RUO
Immunogen Recombinant human GM-CSF protein
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Clone BVD2-21C11
Cross Reactivity Human
Cross Reactivity Primate
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1437
Test Specificity Cynomolgus Monkey, Rhesus Monkey.
Concentration 0.2mg/mL
Host Species Rat
Format Purified
Antigen GM-CSF