Please login to your online account to display your discounted pricing

anti-GM-CSF, Clone: BVD2-21C11, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone BVD2-21C11
Host Species Rat
Isotype κ
Immunogen Recombinant human GM-CSF protein
Antigen GM-CSF
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232967

 
novus biologicals canada
NBP232967
Unlabeled 0.5mL Each for $321.00

NBP2329671M

 
novus biologicals canada
NBP2329671ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone BVD2-21C11
Host Species Rat
Isotype κ
Immunogen Recombinant human GM-CSF protein
Antigen GM-CSF
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Cynomolgus Monkey, Rhesus Monkey.
Gene Accession No. P04141
Gene ID (Entrez) 1437
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL