Please login to your online account to display your discounted pricing

anti-GM-CSF, Clone: BVD2-21C11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329671ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329671M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rat
Immunogen Recombinant human GM-CSF protein
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 1437
Test Specificity Cynomolgus Monkey, Rhesus Monkey.
Clone BVD2-21C11
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P04141
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Antigen GM-CSF
Format Purified