Please login to your online account to display your discounted pricing

anti-GITR/TNFRSF18, Clone: DTA-1, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone DTA-1
Host Species Rat
Isotype λ
Immunogen Mouse CD25+ CD4+ T cell line NP_683700
Antigen GITR/TNFRSF18
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP226661

 
novus biologicals canada
NBP226661
Unlabeled 0.1mg Each for $307.00

NBP226661SS

 
novus biologicals canada
NBP226661SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone DTA-1
Host Species Rat
Isotype λ
Immunogen Mouse CD25+ CD4+ T cell line NP_683700
Antigen GITR/TNFRSF18
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Gene Accession No. Q9Y5U5
Gene ID (Entrez) 8784
Formulation PBS containing 0.05% BSA.