Please login to your online account to display your discounted pricing

anti-FOLR4, Clone: TH6, Novus Biologicals™

$142.00 - $146.24

Specifications

Clone TH6
Host Species Rat
Isotype κ
Immunogen CD3 mAb-stimulated CD4/25 T cells
Antigen FOLR4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP226651

 
novus biologicals canada
NBP226651
Unlabeled 0.05mg Each for $146.24

NBP226651SS

 
novus biologicals canada
NBP226651SS
Unlabeled 0.025mg Each for $142.00
Description & Specifications

Specifications

Clone TH6
Host Species Rat
Isotype κ
Immunogen CD3 mAb-stimulated CD4/25 T cells
Antigen FOLR4
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene ID (Entrez) 390243
Formulation PBS containing 0.05% BSA.