Please login to your online account to display your discounted pricing

anti-CD34, Clone: MEC 14.7, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001071

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001071

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone MEC 14.7
Immunogen murine transformed endothelioma cell line t-end.
Cross Reactivity Mouse
Antigen CD34
Host Species Rat
Isotype IgG2a
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Quantity 0.1mL
Format Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 947
Concentration 1mg/mL
Primary or Secondary Primary