Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: MEC13.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223604SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223604SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5175
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen mouse endothelial cell line T-end
Cross Reactivity Mouse
Isotype κ
Quantity 1mL
Regulatory Status RUO
Host Species Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Antigen CD31/PECAM-1
Format Supernatant
Clone MEC13.3
Monoclonal or Polyclonal Monoclonal