Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: MEC13.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223604

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223604

  • / Each
 

Description & Specifications

Specifications

Antigen CD31/PECAM-1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Clone MEC13.3
Conjugate Unlabeled
Cross Reactivity Mouse
Format Supernatant
Host Species Rat
Immunogen mouse endothelial cell line T-end
Isotype κ
Quantity 5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5175
Target Species Murine