Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: MEC13.3, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone MEC13.3
Host Species Rat
Isotype IgG2a, κ
Immunogen mouse endothelial cell line T-end
Antigen CD31/PECAM-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001475SS

 
novus biologicals canada
NB6001475SS
Unlabeled 0.025mL Each for $139.00

NB6001475

 
novus biologicals canada
NB6001475
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone MEC13.3
Host Species Rat
Isotype IgG2a, κ
Immunogen mouse endothelial cell line T-end
Antigen CD31/PECAM-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications In vitro Assay
Applications In vivo Assay
Gene ID (Entrez) 5175
Formulation Tris-glycine, 150mM NaCl
Concentration 1.0mg/mL