Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: MEC13.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001475

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001475

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications In vitro Assay
Applications In vivo assay
Immunogen mouse endothelial cell line T-end
Clone MEC13.3
Format Purified
Host Species Rat
Cross Reactivity Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 0.1mL
Antigen CD31/PECAM-1
Regulatory Status RUO
Isotype IgG2a, κ
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 5175
Concentration 1.0mg/mL