Please login to your online account to display your discounted pricing

anti-CD31/PECAM-1, Clone: MEC13.3, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone MEC13.3
Host Species Rat
Isotype κ
Immunogen mouse endothelial cell line T-end
Antigen CD31/PECAM-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223604

 
novus biologicals canada
NBP223604
Unlabeled 5mL Each for $309.05

NBP223604SS

 
novus biologicals canada
NBP223604SS
Unlabeled 1mL Each for $109.00
Description & Specifications

Specifications

Clone MEC13.3
Host Species Rat
Isotype κ
Immunogen mouse endothelial cell line T-end
Antigen CD31/PECAM-1
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Gene ID (Entrez) 5175