Please login to your online account to display your discounted pricing

anti-CD25/IL-2 R alpha, Clone: PC61, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227425SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227425SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Quantity 0.025mg
Host Species Rat
Immunogen The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody.
Antigen CD25/IL-2 R alpha
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene Accession No. P01589
Formulation CD25 antibody at 1mg/mL in sterile PBS. Sodium azide free.
Gene ID (Entrez) 3559
Format Purified
Regulatory Status RUO
Applications Flow Cytometry
Clone PC61
Cross Reactivity Mouse