Please login to your online account to display your discounted pricing

anti-CD25/IL-2 R alpha, Clone: PC61, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227425

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227425

  • / Each
 

Description & Specifications

Specifications

Formulation CD25 antibody at 1mg/mL in sterile PBS. Sodium azide free.
Immunogen The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody.
Quantity 0.1mg
Cross Reactivity Mouse
Applications Flow Cytometry
Gene ID (Entrez) 3559
Gene Accession No. P01589
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Concentration 1mg/mL
Regulatory Status RUO
Conjugate Unlabeled
Clone PC61
Antigen CD25/IL-2 R alpha
Host Species Rat
Format Purified