Please login to your online account to display your discounted pricing

anti-CD25/IL-2 R alpha, Clone: PC61, Novus Biologicals™

$109.00 - $252.00

Specifications

Clone PC61
Host Species Rat
Immunogen The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody.
Antigen CD25/IL-2 R alpha
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227425

 
novus biologicals canada
NBP227425
Unlabeled 0.1mg Each for $252.00

NBP227425SS

 
novus biologicals canada
NBP227425SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone PC61
Host Species Rat
Immunogen The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody.
Antigen CD25/IL-2 R alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Gene Accession No. P01589
Gene ID (Entrez) 3559
Formulation CD25 antibody at 1mg/mL in sterile PBS. Sodium azide free.
Concentration 1mg/mL