Please login to your online account to display your discounted pricing

anti-CCR6, Clone: 18B9E6, Novus Biologicals™

$139.00 - $219.00

Specifications

Clone 18B9E6
Host Species Rat
Isotype κ
Immunogen A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody.
Antigen CCR6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225220

 
novus biologicals canada
NBP225220
Unlabeled 0.1mg Each for $219.00

NBP225220SS

 
novus biologicals canada
NBP225220SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 18B9E6
Host Species Rat
Isotype κ
Immunogen A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody.
Antigen CCR6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This product reacts with Mouse.
Gene ID (Entrez) 1235
Formulation CD196 (CCR6) antibody at 0.5mg/mL in PBS, 0.05% BSA
Concentration 0.5mg/mL