Please login to your online account to display your discounted pricing

anti-CCL20/MIP-3 alpha, Clone: 4N8C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227143

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227143

  • / Each
 

Description & Specifications

Specifications

Antigen CCL20/MIP-3 alpha
Applications Western Blot
Clone 4N8C7
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rat
Immunogen Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6364
Test Specificity This monoclonal antibody is specific to Mouse CCL20, it will not cross-react with Human CCL20.
Target Species Human