Please login to your online account to display your discounted pricing

anti-CCL20/MIP-3 alpha, Clone: 4N8C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227143SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227143SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Applications Western Blotting
Gene ID (Entrez) 6364
Antigen CCL20/MIP-3 alpha
Immunogen Full length recombinant mouse CCL20 (MIP-3a) was used as immunogen for the antibody. full length protein
Cross Reactivity Human
Format Purified
Host Species Rat
Quantity 0.025mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity This monoclonal antibody is specific to Mouse CCL20, it will not cross-react with Human CCL20.
Clone 4N8C7
Primary or Secondary Primary