Please login to your online account to display your discounted pricing

anti-Caspase 11, Clone: 17D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12010454

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12010454

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Cross Reactivity Mouse
Quantity 0.1mL
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Microarray
Applications Western Blotting
Concentration 1.0mg/mL
Format Purified
Clone 17D9
Monoclonal or Polyclonal Monoclonal
Antigen Caspase 11
Immunogen A recombinant p30 subunit of mouse Caspase 11. [UniProt No. P70343]
Primary or Secondary Primary
Gene ID (Entrez) 9169
Conjugate Unlabeled
Isotype IgG2a
Formulation PBS