Please login to your online account to display your discounted pricing

anti-Caspase 11, Clone: 17D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12010454SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12010454T

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Microarray
Applications Western Blotting
Quantity 0.025mL
Format Purified
Immunogen A recombinant p30 subunit of mouse Caspase 11. [UniProt No. P70343]
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9169
Isotype IgG2a
Cross Reactivity Mouse
Host Species Rat
Monoclonal or Polyclonal Monoclonal
Clone 17D9
Conjugate Unlabeled
Antigen Caspase 11
Formulation PBS
Concentration 1.0mg/mL