missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RAS-like, family 12, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RASL12 protein.
Supplier: Abnova Corporation H00051285B02
Description
Sequence: MSSVFGKPRAGSGPQSAPLEVNLAILGRRGAGKSALTVKFLTKRFISEYDPNLEDTYSSEETVDHQPVHLRVMDTADLDTPRNCERYLNWAHAFLVVYSVDSRQSFDSSSSYLELLALHAKETQRSIPALLLGNKLDMAQYRQVTKAEGVALAGRFGCLFFEVSACLDFEHVQHVFHEAVREARRELEKSPLTRPLFISEERALPHQAPLTARHGLASCTFNTLSTINLKEMPTVAQAKLVTVKSSRAQSKRKAPTLTLLKGFKIFSpecifications
RAS-like, family 12 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human RASL12 protein. | |
RASL12 | |
RIS | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_016563 | |
RASL12 | |
RASL12 (NP_057647, 1 a.a. ∼ 266 a.a) full-length human protein. | |
RUO | |
Yes | |
51285 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |