Please login to your online account to display your discounted pricing

anti-Rap2A, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 140-183 of human RAP2A was used as the immunogen for the antibody.
Antigen Rap2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224574

 
novus biologicals canada
NBP224574
Unlabeled 0.1mg Each for $257.41

NBP224574SS

 
novus biologicals canada
NBP224574SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 140-183 of human RAP2A was used as the immunogen for the antibody.
Antigen Rap2A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The rat amino acid sequence used as immunogen is 100% homologous in rat and mouse and 94% homologous in human.
Gene Accession No. P10114
Gene ID (Entrez) 5911
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL