Please login to your online account to display your discounted pricing

anti-RAMP2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 68-99 of human RAMP2 were used as the immunogen for this antibody.
Antigen RAMP2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223672

 
novus biologicals canada
NBP223672
Unlabeled 0.1mL Each for $257.41

NBP223672SS

 
novus biologicals canada
NBP223672SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 68-99 of human RAMP2 were used as the immunogen for this antibody.
Antigen RAMP2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 95% homologous in dog and chimpanzee, 90% homologous in pig and red deer, and 85% in buffalo and goat. IL-35 is a heterodimeric protein of p35/IL-12A and EBI3, with a molecular weight of approximately 78 kD. The recombinant IL-35 used for testing also contains a human Fc tag resulting in the approximately 100 kD size seen.
Gene Accession No. O60895
Gene ID (Entrez) 10266
Formulation 100μL of antibody; rabbit sera with 50% glycerol