Please login to your online account to display your discounted pricing

anti-RAMP2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223672

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223672

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 95% homologous in dog and chimpanzee, 90% homologous in pig and red deer, and 85% in buffalo and goat. IL-35 is a heterodimeric protein of p35/IL-12A and EBI3, with a molecular weight of approximately 78 kD. The recombinant IL-35 used for testing also contains a human Fc tag resulting in the approximately 100 kD size seen.
Formulation 100μL of antibody; rabbit sera with 50% glycerol
Applications Western Blotting
Quantity 0.1mL
Gene ID (Entrez) 10266
Cross Reactivity Human
Format Antiserum
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Gene Accession No. O60895
Host Species Rabbit
Regulatory Status RUO
Immunogen Within the range of amino acids 68-99 of human RAMP2 were used as the immunogen for this antibody.
Antigen RAMP2