missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-RAGE, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350-400 of human RAGE was used as the immunogen.
Antigen RAGE
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP203951 Novus Biologicals Canada
NBP203951
0.1mg Each for $307.00
NBP203951SS Novus Biologicals Canada
NBP203951SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
RAGE
Affinity Purified
RUO
Note: This antibody was raised against an amino acid sequence that is 100% conserved in human, however western blot testing shows that it does not detected the expected ∽45 kD band in human lung lysate. Reacts with Mouse and Rat. Predicted to react with Human and New World Monkey.
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acid 350-400 of human RAGE was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
5891