Please login to your online account to display your discounted pricing

anti-RAGE, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human RAGE was used as the immunogen.
Antigen RAGE
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203950SS

 
novus biologicals canada
NBP203950SS
Unlabeled 0.025mg Each for $139.00

NBP203950

 
novus biologicals canada
NBP203950
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human RAGE was used as the immunogen.
Antigen RAGE
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 5891
Formulation 100μg in 200μL PBS containing 0.05% BSA