Please login to your online account to display your discounted pricing

anti-RAGE, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203951SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203951SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5891
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity Note: This antibody was raised against an amino acid sequence that is 100% conserved in human, however western blot testing shows that it does not detected the expected ∽45 kD band in human lung lysate. Reacts with Mouse and Rat. Predicted to react with Human and New World Monkey.
Format Affinity Purified
Conjugate Unlabeled
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Immunogen A portion of amino acid 350-400 of human RAGE was used as the immunogen.
Quantity 0.025mg
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen RAGE