Please login to your online account to display your discounted pricing

anti-RAGE, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203950

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203950

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 5891
Immunogen A portion of amino acids 1-60 of human RAGE was used as the immunogen.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Quantity 0.1mg
Antigen RAGE
Host Species Rabbit
Format Purified