Please login to your online account to display your discounted pricing

anti-Rad9, Polyclonal, Novus Biologicals™

$182.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Fusion protein of full length human Rad9 protein. [UniProt No. Q99638]
Antigen Rad9
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100193

 
novus biologicals canada
NB100193
Unlabeled 0.05mL Each for $309.05

NB100193SS

 
novus biologicals canada
NB100193SS
Unlabeled 0.025mL Each for $182.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Fusion protein of full length human Rad9 protein. [UniProt No. Q99638]
Antigen Rad9
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q99638
Gene ID (Entrez) 5883
Formulation Tris-glycine, 150mM NaCl