Please login to your online account to display your discounted pricing

anti-Rad51D, Polyclonal, Novus Biologicals™

$144.63 - $329.00

Specifications

Host Species Rabbit
Immunogen A peptide from human Rad51D (homologue 4). [UniProt No. O75771]
Antigen Rad51D
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100166

 
novus biologicals canada
NB100166
Unlabeled 0.1mL Each for $329.00

NB100166SS

 
novus biologicals canada
NB100166SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide from human Rad51D (homologue 4). [UniProt No. O75771]
Antigen Rad51D
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This is specific for hRAD51D. There is no cross-reactivity observed with XRCC2, XRCC3, RAD51, RAD51B, or RAD51C. Human. This antibody does not work in mouse.
Gene Accession No. O75771
Gene ID (Entrez) 5892
Formulation PBS
Concentration 1.9mg/mL