Please login to your online account to display your discounted pricing

anti-Rad51D, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100166SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100166SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 1.9mg/mL
Conjugate Unlabeled
Formulation PBS
Gene ID (Entrez) 5892
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This is specific for hRAD51D. There is no cross-reactivity observed with XRCC2, XRCC3, RAD51, RAD51B, or RAD51C. Human. This antibody does not work in mouse.
Gene Accession No. O75771
Host Species Rabbit
Antigen Rad51D
Format Affinity Purified
Immunogen A peptide from human Rad51D (homologue 4). [UniProt No. O75771]
Regulatory Status RUO