Please login to your online account to display your discounted pricing

anti-Rad51D, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100166

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100166

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Format Affinity Purified
Formulation PBS
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Concentration 1.9mg/mL
Quantity 0.1mL
Regulatory Status RUO
Conjugate Unlabeled
Gene ID (Entrez) 5892
Gene Accession No. O75771
Monoclonal or Polyclonal Polyclonal
Test Specificity This is specific for hRAD51D. There is no cross-reactivity observed with XRCC2, XRCC3, RAD51, RAD51B, or RAD51C. Human. This antibody does not work in mouse.
Antigen Rad51D
Cross Reactivity Human
Immunogen A peptide from human Rad51D (homologue 4). [UniProt No. O75771]
Host Species Rabbit