Please login to your online account to display your discounted pricing

anti-Rad50, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001487

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001487

  • / Each
 

Description & Specifications

Specifications

Antigen Rad50
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Gene Accession No. Q92878
Host Species Rabbit
Immunogen Human RAD50 zinc hook region (amino acids 628-787) expressed in E. coli. [UniProt No. Q92878]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10111
Target Species Human