missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Rad17, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen This antibody was raised against RAD17
Antigen Rad17
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227106 Novus Biologicals Canada
NBP227106
0.1mg Each for $257.41
NBP227106SS Novus Biologicals Canada
NBP227106SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Rad17
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat.
5884
0.5mg/mL
This antibody was raised against RAD17
Polyclonal
Primary
Western Blot
O75943
100μg in 200μL PBS containing 0.05% BSA.