Please login to your online account to display your discounted pricing

anti-Rad17, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227106

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227106

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA.
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5884
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat.
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Antigen Rad17
Gene Accession No. O75943
Regulatory Status RUO
Format Affinity Purified
Conjugate Unlabeled
Host Species Rabbit
Applications Western Blotting
Immunogen This antibody was raised against RAD17