missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Rad17 (p Ser647), Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acids 644-652 (LPLBQNSGS) of mouse Rad17. The B represents a phosphorylated serine residue.
Antigen Rad17 (p Ser647)
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056386T Novus Biologicals Canada
NB10056386SS
0.025mg Each for $139.00
NB10056386 Novus Biologicals Canada
NB10056386
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
Rad17 (p Ser647)
Purified
RUO
RAD17 Phosphospecific [Ser647]. Cross reacts with Mouse. Predicted to react with Human and Rat.
5884
0.5mg/mL
The antibody was raised against a synthetic peptide corresponding to the amino acids 644-652 (LPLBQNSGS) of mouse Rad17. The B represents a phosphorylated serine residue.
Polyclonal
Primary
Western Blot
O75943
PBS containing 0.2% gelatin