Please login to your online account to display your discounted pricing

anti-Rad17 (p Ser647), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056386SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056386T

  • / Each
 

Description & Specifications

Specifications

Antigen Rad17 (p Ser647)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. O75943
Host Species Rabbit
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acids 644-652 (LPLBQNSGS) of mouse Rad17. The B represents a phosphorylated serine residue.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5884
Test Specificity RAD17 Phosphospecific [Ser647]. Cross reacts with Mouse. Predicted to react with Human and Rat.
Target Species Murine