Please login to your online account to display your discounted pricing

anti-Rad1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056320SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056320T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Gene Accession No. O60671
Applications Western Blotting
Gene ID (Entrez) 5810
Concentration 0.5mg/mL
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acid 133-148 of human Rad1 (accession number O60671)
Quantity 0.025mg
Antigen Rad1
Conjugate Unlabeled
Regulatory Status RUO
Host Species Rabbit
Cross Reactivity Human
Format Purified