Please login to your online account to display your discounted pricing

anti-Rad1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056320

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056320

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O60671
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acid 133-148 of human Rad1 (accession number O60671)
Primary or Secondary Primary
Conjugate Unlabeled
Applications Western Blotting
Cross Reactivity Human
Quantity 0.1mg
Gene ID (Entrez) 5810
Host Species Rabbit
Regulatory Status RUO
Antigen Rad1
Format Purified